SY3H3D67D2BA Synthonix Corporation

Ethyl 3-bromo-1-methyl-1H-pyrazole-4-carboxylate

  • Cas Number : 139308-52-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/46313.html

Safety Information

MSDS https://synthonix.com/46313.html
Availability details for
Quantity
  • Availability
  • Add to Cart