SY3H3D67D2C4 Synthonix Corporation

Ethyl 2-amino-4-hydroxypyrimidine-5-carboxylate

  • Cas Number : 15400-53-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/46353.html

Safety Information

MSDS https://synthonix.com/46353.html
Availability details for
Quantity
  • Availability
  • Add to Cart