SY3H3D67D2CE Synthonix Corporation

4-Ethyl-2-methylthiazole-5-carboxylic acid

  • MDL number MFCD12027007
  • Cas Number : 119778-44-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/46419.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/46419.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart