SY3H3D67D2DA Synthonix Corporation

Ethyl 5-aminovalerate hydrochloride

  • MDL number MFCD12546205
  • Cas Number : 29840-57-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form White to off-white powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/46517.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/46517.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart