SY3H3D67D2DB Synthonix Corporation

(R)-Ethyl 2-amino-3-methylbutanoate hydrochloride

  • MDL number MFCD00191043
  • Cas Number : 73913-64-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form White to off-white powder or crytals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/46519.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/46519.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart