SY3H3D67D2DC Synthonix Corporation

Ethyl 3-amino-2,2-dimethylpropanoate hydrochloride

  • Cas Number : 80253-38-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/46520.html

Safety Information

MSDS https://synthonix.com/46520.html
Availability details for
Quantity
  • Availability
  • Add to Cart