SY3H3D67D2DD Synthonix Corporation

2,6-Diethyl-4-methylaniline

  • MDL number MFCD00035950
  • Cas Number : 24544-08-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Clear liquid / Colour: Very pale yellow - Yellow red
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/54977.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/54977.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart