SY3H3D67D2E6 Synthonix Corporation

4-Ethoxybenzene-1,3-diamine sulfate

  • MDL number MFCD00035493
  • Cas Number : 68015-98-5
  • Molecular Weight : 250.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-ethoxybenzene-1,3-diamine sulfate
Inchi Code 1S/C8H12N2O.H2O4S/c1-2-11-8-4-3-6(9)5-7(8)10;1-5(2,3)4/h3-5H,2,9-10H2,1H3;(H2,1,2,3,4)
Inchi Key WRYXZLYZWDZVKS-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. Refrigerator
Physical Form White to yellow to purple powder or crystals
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/46634.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/46634.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart