SY3H3D67D2E9 Synthonix Corporation

2-Ethyl-5-nitroaniline

  • MDL number MFCD00034056
  • Cas Number : 20191-74-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Crystal - Powder / Colour: Yellow - Deep reddish yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/46659.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/46659.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart