SY3H3D67D2EB Synthonix Corporation

Ethyl 3-aminopicolinate

  • MDL number MFCD00661294
  • Cas Number : 27507-15-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form White to pale-yellow to brown powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/46661.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/46661.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart