SY3H3D67D2ED Synthonix Corporation

Ethyl 3-amino-4-pyridinecarboxylate

  • MDL number MFCD03788270
  • Cas Number : 14208-83-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Crystal - Powder / Colour: Gray - Yellow - Brown
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/46678.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/46678.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart