SY3H3D67D2EF Synthonix Corporation

Ethyl 1-(difluoromethyl)-5-methyl-1H-pyrazole-3-carboxylate

  • MDL number MFCD30471703
  • Cas Number : 1998216-36-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/46689.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/46689.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart