SY3H3D67D2F0 Synthonix Corporation

Ethyl 3-(difluoromethyl)-1-methyl-1H-pyrazole-4-carboxylate

  • MDL number MFCD12022341
  • Cas Number : 141573-95-7
  • Molecular Weight : 204.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/46690.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/46690.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart