SY3H3D67D2F8 Synthonix Corporation

2-Ethoxy-4-fluorophenylboronic acid

  • MDL number MFCD03095001
  • Cas Number : 480438-58-2
  • Molecular Weight : 183.97

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Referigrator
Physical Form Crystal - Powder
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/46846.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/46846.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart