SY3H3D67D2FA Synthonix Corporation

4-Ethoxy-3-fluorophenylboronic acid

  • MDL number MFCD04115667
  • Cas Number : 279263-10-4
  • Molecular Weight : 183.97

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Crystal - Powder / Colour: White - Almost white
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/46848.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/46848.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart