SY3H3D67D305 Synthonix Corporation

Ethyl 2-methyl-1H-pyrrole-3-carboxylate

  • MDL number MFCD00101795
  • Cas Number : 936-12-9
  • Molecular Weight : 153.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Pale-yellow to Yellow-brown to Brown Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/46928.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/46928.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart