SY3H3D67D31B Synthonix Corporation

Ethyl 4-oxotetrahydro-2H-thiopyran-3-carboxylate 1,1-dioxide

  • MDL number MFCD08166525
  • Cas Number : 889946-17-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form Pale-yellow to Yellow-brown Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/47044.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/47044.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart