SY3H3D67D31F Synthonix Corporation

Ethyl 2-methyl-3-oxopyrrolidine-1-carboxylate

  • MDL number MFCD24689559
  • Cas Number : 73193-55-2
  • Molecular Weight : 171.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/47065.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/47065.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart