SY3H3D67D320 Synthonix Corporation

Ethyl hex-2-enoate

  • MDL number No MDL found
  • Cas Number : 27829-72-7
  • Molecular Weight : 142.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/47082.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/47082.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart