SY3H3D67D321 Synthonix Corporation

Ethyl hex-3-enoate

  • MDL number No MDL found
  • Cas Number : 2396-83-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/47083.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/47083.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart