SY3H3D67D322 Synthonix Corporation

Ethyl 2,2-difluorohexanoate

  • Cas Number : 74106-81-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin USA
Product Link https://synthonix.com/47090.html

Safety Information

MSDS https://synthonix.com/47090.html
Availability details for
Quantity
  • Availability
  • Add to Cart