SY3H3D67D325 Synthonix Corporation

Ethyl 2-ethyl-3-oxobutanoate

  • MDL number MFCD00039898
  • Cas Number : 607-97-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Clear liquid / Colour: Colorless - Yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/47095.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/47095.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart