SY3H3D67D34D Synthonix Corporation

Ethyl 3-cyano-1H-pyrrole-2-carboxylate

  • Cas Number : 7126-44-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/48824.html

Safety Information

MSDS https://synthonix.com/48824.html
Availability details for
Quantity
  • Availability
  • Add to Cart