SY3H3D67D354 Synthonix Corporation

Ethyl 2-bromoisonicotinate

  • MDL number MFCD06205252
  • Cas Number : 89978-52-9
  • Molecular Weight : 230.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form White to yellow or light-red powder or crystals or liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/48906.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/48906.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart