SY3H3D67D35A Synthonix Corporation

Ethyl 2-Chloropyridine-4-carboxylate

  • MDL number MFCD04114259
  • Cas Number : 54453-93-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Colorless to Pale-yellow Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/48953.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/48953.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart