SY3H3D67D360 Synthonix Corporation

Ethyl 4-oxo-4H-pyran-2-carboxylate

  • Cas Number : 1551-45-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/49117.html

Safety Information

MSDS https://synthonix.com/49117.html
Availability details for
Quantity
  • Availability
  • Add to Cart