SY3H3D67D362 Synthonix Corporation

1-Ethyl-4-iodobenzene

  • MDL number MFCD00060663
  • Cas Number : 25309-64-2
  • Molecular Weight : 232.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Referigrator
Physical Form Colorless to Yellow to Red-brown to Brown Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/49167.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/49167.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart