SY3H3D67D366 Synthonix Corporation

Ethyl 5-bromo-2-(methylthio)pyrimidine-4-carboxylate

  • Cas Number : 74840-38-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/49182.html

Safety Information

MSDS https://synthonix.com/49182.html
Availability details for
Quantity
  • Availability
  • Add to Cart