SY3H3D67D379 Synthonix Corporation

5-Ethoxypicolinic acid

  • Cas Number : 98353-08-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/49300.html

Safety Information

MSDS https://synthonix.com/49300.html
Availability details for
Quantity
  • Availability
  • Add to Cart