SY3H3D67D37D Synthonix Corporation

Ethyl 6-hydroxynicotinate

  • MDL number MFCD08460993
  • Cas Number : 18617-50-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form White to Gray to Yellow Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/49308.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/49308.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart