SY3H3D67D38C Synthonix Corporation

N,N-Dimethyl-2-(p-tolyloxy)ethanamine

  • MDL number MFCD06304626
  • Cas Number : 51344-14-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/54992.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/54992.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart