SY3H3D67D398 Synthonix Corporation

Ethyl 4-amino-7H-pyrrolo[2,3-d]pyrimidine-5-carboxylate

  • Cas Number : 1696907-15-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/49760.html

Safety Information

MSDS https://synthonix.com/49760.html
Availability details for
Quantity
  • Availability
  • Add to Cart