SY3H3D67D788 Synthonix Corporation

Ethyl 2-amino-2-(3-chloropyridin-4-yl)acetate hydrochloride

  • MDL number MFCD30186146
  • Cas Number : 1956319-58-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerator
Physical Form Yellow solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/49987.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/49987.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart