SY3H3D67D79F Synthonix Corporation

4-Ethyl-1-iodo-2-methylbenzene

  • MDL number MFCD27996329
  • Cas Number : 866996-02-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerator
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/49973.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/49973.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart