SY3H3D67D7A2 Synthonix Corporation

Ethyl 2-amino-6-chlorobenzoate

  • Cas Number : 172217-11-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/49805.html

Safety Information

MSDS https://synthonix.com/49805.html
Availability details for
Quantity
  • Availability
  • Add to Cart