SY3H3D67D7AF Synthonix Corporation

Ethyl 4-acetyl-1H-pyrrole-2-carboxylate

  • MDL number MFCD09263957
  • Cas Number : 119647-69-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/50096.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/50096.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart