SY3H3D67D7B0 Synthonix Corporation

Ethyl 3-oxo-3-(1H-pyrrol-2-yl)propanoate

  • Cas Number : 169376-35-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/50098.html

Safety Information

MSDS https://synthonix.com/50098.html
Availability details for
Quantity
  • Availability
  • Add to Cart