SY3H3D67D7B3 Synthonix Corporation

Ethyl 4-hydroxy-6-methyl-2-oxo-1,2-dihydropyridine-3-carboxylate

  • MDL number MFCD00205625
  • Cas Number : 10350-10-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form White to light-yellow to yellow powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/50111.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/50111.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart