SY3H3D67D7B5 Synthonix Corporation

2-Ethoxybenzohydrazide

  • MDL number MFCD00017050
  • Cas Number : 21018-13-3
  • Molecular Weight : 180.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-ethoxybenzohydrazide
Inchi Code 1S/C9H12N2O2/c1-2-13-8-6-4-3-5-7(8)9(12)11-10/h3-6H,2,10H2,1H3,(H,11,12)
Inchi Key LADUENYEZHMRQH-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/50176.html

Safety Information

signalword Warning
pcodes P261,P305,P351,P338
MSDS https://synthonix.com/50176.html
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart