SY3H3D67D7C7 Synthonix Corporation

5-Ethyl-2-methoxyaniline

  • MDL number MFCD00666677
  • Cas Number : 67291-61-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerator
Physical Form White to off-white powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/50338.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/50338.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart