SY3H3D67D7C8 Synthonix Corporation

(4-Ethoxyphenyl)methanamine

  • MDL number MFCD02225920
  • Cas Number : 6850-60-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form White to yellow powder or crystals or semi-solid or liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/50343.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/50343.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart