SY3H3D67D7CE Synthonix Corporation

Ethyl 5-Propylpyrazole-3-carboxylate

  • MDL number MFCD08700607
  • Cas Number : 92945-27-2
  • Molecular Weight : 182.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/50402.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/50402.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart