SY3H3D67D7D3 Synthonix Corporation

Ethyl 2-(2-oxocyclopentyl)acetate

  • MDL number MFCD00044715
  • Cas Number : 20826-94-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Clear liquid / Colour: Colorless - Yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/50438.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/50438.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart