SY3H3D67D7D6 Synthonix Corporation

Ethyl 2-(ethoxymethylene)-3-oxobutanoate

  • MDL number MFCD06797311
  • Cas Number : 3788-94-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form Clear liquid / Colour: Colorless - Yellow - Reddish yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/50445.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/50445.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart