SY3H3D67D7D8 Synthonix Corporation

Ethyl 3-oxo-3-(tetrahydrofuran-2-yl)propanoate

  • MDL number MFCD06410652
  • Cas Number : 887411-85-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form Red-brown Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/50451.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/50451.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart