SY3H3D67D7E0 Synthonix Corporation

trans-4-Ethylcyclohexanecarboxylic acid

  • MDL number MFCD01076323
  • Cas Number : 6833-47-2
  • Molecular Weight : 156.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-ethylcyclohexanecarboxylic acid
Inchi Code 1S/C9H16O2/c1-2-7-3-5-8(6-4-7)9(10)11/h7-8H,2-6H2,1H3,(H,10,11)/t7-,8-
Inchi Key UNROFSAOTBVBBT-ZKCHVHJHSA-N

Description

Purity 98%
Storage temp. Refrigerator
Physical Form Crystal - Powder / Colour: White - Almost white
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/50506.html

Safety Information

signalword Warning
pcodes P261,P305,P351,P338
MSDS https://synthonix.com/50506.html
pictograms GHS07
hstatements H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart