SY3H3D67D7E1 Synthonix Corporation

Ethyl 2-hydroxycyclohexanecarboxylate

  • Cas Number : 3444-72-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/50509.html

Safety Information

MSDS https://synthonix.com/50509.html
Availability details for
Quantity
  • Availability
  • Add to Cart