SY3H3D67D7E5 Synthonix Corporation

7-Ethoxy-7-oxoheptanoic acid

  • MDL number MFCD00014383
  • Cas Number : 33018-91-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form Clear liquid / Colour: Colorless - Yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/50517.html

Safety Information

signalword Warning
MSDS https://synthonix.com/50517.html
pictograms GHS07
hstatements H315, H319
Availability details for
Quantity
  • Availability
  • Add to Cart