SY3H3D67D7E6 Synthonix Corporation

Ethyl 2-((tetrahydro-2H-pyran-2-yl)oxy)acetate

  • MDL number MFCD00128492
  • Cas Number : 61675-94-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/50521.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/50521.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart