SY3H3D67D7E9 Synthonix Corporation

Ethyl 1-methylpipecolinate

  • MDL number MFCD00006492
  • Cas Number : 30727-18-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Clear liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/50530.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/50530.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart